Learn More
Biomatik Corporation Mouse Autophagy Related Protein 5 (ATG5) ELISA Kit
Supplier:  Biomatik Corporation EKU10674
Encompass_Preferred
Mouse Autophagy Related Protein 5 (ATG5) ELISA Kit for detecting ATG5 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99J83. Gene ID: 11793. Target Synonyms: ASP; APG5; APG5L; hAPG5; Apoptosis-specific protein; Autophagy protein 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8313