Learn More
Sino Biological Recombinant Mouse Autotaxin/ENPP-2 Protein (His Tag)
Supplier:  Sino Biological 50663M07H500
Encompass_Preferred
A DNA sequence encoding the mature form of mouse ENPP2 (Q9R1E6-1) (Ser 49-Ile 862) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-7143