Learn More
Sino Biological Mouse AXL Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785262
Encompass_Preferred
The Mouse Axl ELISA Pair Set is for the quantitative determination of Mouse Axl.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse Axl coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse Axl present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse Axl monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse Axl present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85262 Supplier Sino Biological Supplier No. SEK501265
The Mouse Axl ELISA Pair Set is for the quantitative determination of Mouse Axl.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse Axl coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse Axl present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse Axl monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse Axl present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.