Learn More
Sino Biological Anti-AXL Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50126RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse AXL (rM AXL; Catalog#50126-M08H; NP_033491.2; Met 1-Pro 443). AXL specific IgG was purified by mouse AXL affinity chromatography.
Catalog No. 50-241-0625