Learn More
American Research Products Inc Mouse B cell leukemia lymphoma
Catalog No. 5000329647
Encompass
Mouse B-cell leukemia/lymphoma 2 Bcl-2 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29647 Supplier American Research Products Inc Supplier No. CSBE08855M
Mouse B-cell leukemia/lymphoma 2 Bcl-2 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma Method Sandwich