Learn More
American Research Products Inc Mouse b2 microglobulin BMG b2
Catalog No. 5000329047
Encompass
Mouse b2-microglobulin BMG/b2-MG ELISA Kit from CUSABIO Detection Range 0 156 g/ml-10 g/ml Sensitivity 0 156 g/ml Sample serum plasma urine tissue homogenates Method Competitive
Catalog No. 50-003-29047 Supplier American Research Products Inc Supplier No. CSBE13053M
Mouse b2-microglobulin BMG/b2-MG ELISA Kit from CUSABIO Detection Range 0 156 g/ml-10 g/ml Sensitivity 0 156 g/ml Sample serum plasma urine tissue homogenates Method Competitive