Learn More
Sino Biological Anti-Beta-2 microglobulin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50957T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Beta-2 microglobulin (rM Beta-2 microglobulin; Catalog#50957-M08H; P01887; Met1-Met119). Beta-2 microglobulin specific IgG was purified by Mouse Beta-2 microglobulin affinity chromatography.
Catalog No. 50-241-2175