Learn More
Lonza Walkersville Mouse B6C3F1 Heps Cryo Plateab
Supplier:  Lonza Walkersville MXCP01
Encompass
All our plateable cryopreserved primary animal hepatocytes are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension They are tested for healthy morphology lasting 72 hours in monlayer sandwich culture
Catalog No. 50-004-96275