Learn More
Bon Opus Biosciences Recombinant Mouse CD275/B7-H2 (C-mFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP100550UG
Encompass_Preferred
Recombinant Mouse CD275/B7-H2 (C-mFc) is produced by Human Cells. The target gene encoding Thr48-Lys279 is expressed with a C-mFc tag. Accession# Q9JHJ8. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1005
Catalog No. 50-217-2607