Learn More
Sino Biological Recombinant Mouse IL-2R beta/CD122 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50792M08H1
Encompass_Preferred
A DNA sequence encoding the mouse IL2RB (P16297) (Met1-Glu240) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-222-3766