Learn More
Sino Biological Anti-BACE1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50002RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse BACE1 (rM BACE1; Catalog#50002-M08H; NP_035922.4; Met1-Thr457). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3564