Learn More
Sino Biological Recombinant Mouse BACE2 Protein (His Tag)
Supplier:  Sino Biological 51031M08H1
Encompass_Preferred
A DNA sequence encoding the mouse BACE2 (Q9JL18) (Met1-Pro462) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-255-3023