Learn More
ACROBiosystems Mouse BAFF 200ug
Catalog No. 502535481
Encompass_Preferred
Biotinylated Mouse BAFF Protein AvitagHis Tag (BAF-M82Q3) is expressed from human 293 cells (HEK293). It contains AA Ala 127 - Leu 309 (Accession # Q9WU72-1).
Catalog No. 50-253-5481 Supplier ACROBiosystems Supplier No. BAFM82Q3200UG
Biotinylated Mouse BAFF Protein AvitagHis Tag (BAF-M82Q3) is expressed from human 293 cells (HEK293). It contains AA Ala 127 - Leu 309 (Accession # Q9WU72-1).