Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse BAFF ELISA Kit

Catalog No. EEL082
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL082 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL082 Supplier Invitrogen™ Supplier No. EEL082
Add to Cart
Add to Cart

ELISA

The Mouse BAFF/CD257( B-cell Activating Factor) ELISA quantitates BAFF in serum, plasma, and other biological fluids The Mouse BAFF/CD257( B-cell Activating Factor) solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

BAFF is a membrane bound cytokine expressed by B cell lineage cells. The soluble form of the cytokine binds to the receptors TNFRSF13B/TACI, TNFRSF17/BCMA, and TNFRSF13C/BAFFR. BAFF is a potent B cell activator, and important for the proliferation and differentiation of B cells. BAFF is also expressed by T lymphocytes and has been shown to promote T cell activation and survival. Isoforms representing alternatively spliced, and glycosylated variants have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9WU72
Assay Range 62.5-4000 pg/mL
Assay Sensitivity 37.5 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Tumor necrosis factor ligand
Sample Type Plasma, Serum
Target Species Mouse
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias ApoL related ligand TALL-1; b cell-activating factor; B lymphocyte stimulator; BAFF; B-cell-activating factor; B-lymphocyte stimulator; BLYS; CD257; D8Ertd387e; delta BAFF; Delta4 BAFF; dendritic cell-derived TNF-like molecule; DTL; RGD1561519; TALL1; TALL-1; THANK; TNF- and APOL-related leukocyte expressed ligand 1; TNF and ApoL-related leukocyte expressed ligand 1; TNF homolog that activates apoptosis; Tnfsf13b; TNFSF20; TNLG7A; tumor necrosis factor (ligand) superfamily member 13b; tumor necrosis factor (ligand) superfamily, member 13b; tumor necrosis factor (ligand) superfamily, member 20; tumor necrosis factor ligand 7A; tumor necrosis factor ligand superfamily member 13B; Tumor necrosis factor ligand superfamily member 13b, membrane form; Tumor necrosis factor ligand superfamily member 13b, soluble form; tumor necrosis factor superfamily member 13b; tumor necrosis factor-like protein ZTNF4; UNQ401/PRO738; ZTNF4ApoL related ligand TALL-1; b cell-activating factor; B lymphocyte stimulator; BAFF; B-cell-activating factor; B-lymphocyte stimulator; BLYS; CD257; D8Ertd387e; delta BAFF; Delta4 BAFF; dendritic cell-derived TNF-like molecule; DTL; RGD1561519; TALL1; TALL-1; THANK; TNF- and APOL-related leukocyte expressed ligand 1; TNF and ApoL-related leukocyte expressed ligand 1; TNF homolog that activates apoptosis; Tnfsf13b; TNFSF20; TNLG7A; tumor necrosis factor (ligand) superfamily member 13b; tumor necrosis factor (ligand) superfamily, member 13b; tumor necrosis factor (ligand) superfamily, member 20; tumor necrosis factor ligand 7A; tumor necrosis factor ligand superfamily member 13B; Tumor necrosis factor ligand superfamily member 13b, membrane form; Tumor necrosis factor ligand superfamily member 13b, soluble form; tumor necrosis factor superfamily member 13b; tumor necrosis factor-like protein ZTNF4; UNQ401/PRO738; ZTNF4;
Gene ID (Entrez) 24099
Gene Symbol TNFSF13B
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 100 μL, Serum 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target BAFF (BLyS)
Test Time 1 hr 20 min
Total Assay Time 3 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.