Learn More
BIOHIPPO Mouse BALP Microsample
SDP
Catalog No. 5000315860
Encompass
Mouse BALP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BALP in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 068 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse BALP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse BALP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse BALP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15860 Supplier BIOHIPPO Supplier No. BHE15211942
Mouse BALP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BALP in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 068 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse BALP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse BALP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse BALP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color