Learn More
American Research Products Inc Mouse basic fibroblast growth
Catalog No. 5000326506
Encompass
Mouse basic fibroblast growth factor bFGF ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26506 Supplier American Research Products Inc Supplier No. CSBE08001M
Mouse basic fibroblast growth factor bFGF ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich