Learn More
Sino Biological Anti-BAX Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106076T38100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse BAX, and purified by antigen affinity chromatography.
Catalog No. 50-240-7902