Learn More
Bioworld MOUSE BAX (RECOMBINANT)
Supplier:  Bioworld 220605701
Encompass_Preferred
Recombinant Mouse BAX Source: E. Coli Description: Recombinant Mouse Bax, amino acids 38-171 , produced in E.Coli, is a single, non-glycosylated polypeptide chain. Recombinant Mouse Bax is purified by proprietary chromatographic tech...
Catalog No. 50-253-670