Learn More
Biomatik Corporation Mouse B-Cell CLL/Lymphoma 9 (Bcl9) ELISA Kit
Supplier:  Biomatik Corporation EKU02671
Encompass_Preferred
Mouse B-Cell CLL/Lymphoma 9 (Bcl9) ELISA Kit for detecting Bcl9 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.12-5000pg/ml. Sensitivity: 27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9D219. Gene ID: 77578. Target Synonyms: LGS; Protein legless homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9103