Learn More
Sino Biological Mouse BCHE Protein 50ug (Sino Biological)
Supplier:  Sino Biological 50418M08H50
Encompass_Preferred
A DNA sequence encoding the mouse BCHE (NP_033868.3) (Met 1-Leu 603) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-237-2252