Learn More
Raybiotech Inc Mouse BCL-W ELISA Kit
Supplier:  Raybiotech Inc ELMBCLW1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.41 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse BCL-W.
Catalog No. 50-194-9929