Learn More
ACROBiosystems ACROBiosystems Mouse BCMA / TNFRSF17 Protein, Fc Tag
Supplier:  ACROBiosystems BCAM5258100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse BCMA, Fc Tag (BCA-M5258) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Thr 49 (Accession # O88472-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 32.1 kDa. The protein migrates as 36 kDa under reducing (R) condition (SDS-PAGE) due to O-linked glycosylation which has been verified by O-Glycosidase and Neuraminidase digestion.;Immobilized Mouse BAFF, Mouse IgG2a Fc Tag (Cat. No. BAF-M5257) at 5 μg/mL (100 μL/well) can bind Mouse BCMA, Fc Tag (Cat. No. BCA-M5258) with a linear range of 2-20 ng/mL (QC tested).;Immobilized Human APRIL, His,Flag Tag, active trimer (Cat. No. APL-H52D1) at 2 μg/mL (100 μL/well) can bind Mouse BCMA, Fc Tag (Cat. No. BCA-M5258) with a linear range of 0.3-2 ng/mL (Routinely tested).
Catalog No. 50-210-6456