Learn More
American Research Products Inc Mouse Beta Endorphin b EP ELI
Catalog No. 5000330198
Encompass
Mouse Beta-Endorphin b-EP ELISA Kit from CUSABIO Detection Range 0 78 pg/mL-50 pg/mL Sensitivity 0 195 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30198 Supplier American Research Products Inc Supplier No. CSBE06827M
Mouse Beta-Endorphin b-EP ELISA Kit from CUSABIO Detection Range 0 78 pg/mL-50 pg/mL Sensitivity 0 195 pg/mL Sample serum plasma tissue homogenates Method Sandwich