Learn More
Abcam Mouse beta IGH3 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1937171X96TESTS
Encompass_Preferred
Mouse beta IGH3 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse beta IGH3 in Mouse in Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2316160