Learn More
ACROBiosystems Mouse Beta-NGF 20ug
Catalog No. 502535472
Encompass_Preferred
Mouse Beta-NGF Protein Tag Free (BEF-M5213) is expressed from human 293 cells (HEK293). It contains AA Ser 122 - Gly 241 (Accession # P01139).
Catalog No. 50-253-5472 Supplier ACROBiosystems Supplier No. BEFM521320UG
Mouse Beta-NGF Protein Tag Free (BEF-M5213) is expressed from human 293 cells (HEK293). It contains AA Ser 122 - Gly 241 (Accession # P01139).