Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse Betacellulin Recombinant Protein, PeproTech®

Catalog No. 315215UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
315-21-5UG 5 μg
31521100UG 100 μg
315-21-1MG 1 mg
315-21-20UG 20 μg
31521250UG 250 μg
31521500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 315-21-5UG Supplier Gibco™ Supplier No. 315215UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine Betacellulin is a 9.0 kDa monomeric protein, containing 80 amino acid residues, which comprises the mature EGF-homologous portion of the Betacellulin protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q05928
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 12223
Molecular Weight (g/mol) 9 kDa
Name Mouse Betacellulin
Purification Method Purified
Quantity 5 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias Bcn; betacellulin; betacellulin, epidermal growth factor family member; BTC; OTTHUMP00000160600; OTTHUMP00000219057; Probetacellulin
Common Name Betacellulin
Gene Symbol BTC
Biological Activity Determined by its ability to stimulate the proliferation of mouse Balb/3T3 cells. The expected ED50 is ≤ 0.01 ng/ml, corresponding to a specific activity of ≥ 1 x 10^8 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence DGNTTRTPET NGSLCGAPGE NCTGTTPRQK VKTHFSRCPK QYKHYCIHGR CRFVVDEQTP SCICEKGYFG ARCERVDLFY
Content And Storage -20°C
Activity Determined by its ability to stimulate the proliferation of mouse Balb/3T3 cells. The expected ED50 is ≤ 0.01 ng/ml, corresponding to a specific activity of ≥ 1 x 10^8 units/mg.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.