Learn More
Biomatik Corporation Mouse Betaine Homocysteine Methyltransferase (BHMT) ELISA Kit
Supplier:  Biomatik Corporation EKU12459
Encompass_Preferred
Mouse Betaine Homocysteine Methyltransferase (BHMT) ELISA Kit for detecting BHMT in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35490. Gene ID: 12116. Target Synonyms: Betaine—homocysteine S-methyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7962