Learn More
Crystal Chem Inc MOUSE BILE ACIDS ASSAY,96T
SDP
Catalog No. NC2043612
Encompass
Mouse Total Bile Acids Assay Kit (New Version)The Mouse Total Bile Acids Assay is used to quantify levels of total bile acids in mouse serum and plasma in 10 minutes.
Catalog No. NC2043612 Supplier Crystal Chem Inc Supplier No. 80471
Mouse Total Bile Acids Assay Kit (New Version)The Mouse Total Bile Acids Assay is used to quantify levels of total bile acids in mouse serum and plasma in 10 minutes.