Learn More
Sino Biological Anti-BLMH Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50668RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse BLMH / Bleomycin Hydrolase (rM BLMH / Bleomycin Hydrolase; Catalog#50668-M07E; NP_848760.1; Asn 2-Glu 455). BLMH / Bleomycin Hydrolase specific IgG was purified by Mouse BLMH / Bleomycin Hydrolase affinity chromatography.
Catalog No. 50-240-8974