Learn More
Sino Biological Mouse BLMH Protein 50ug (Sino Biological)
Supplier:  Sino Biological 50668M07E50
Encompass_Preferred
A DNA sequence encoding the mouse BLMH (NP_848760.1) (Asn 2-Glu 455) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-237-2564