Learn More
Biomatik Corporation Mouse B-Lymphocyte Chemoattractant (BLC1) CLIA Kit
Supplier:  Biomatik Corporation EKU09678
Encompass_Preferred
Mouse B-Lymphocyte Chemoattractant (BLC1) CLIA Kit for detecting BLC1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.52pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O55038. Gene ID: 55985. Target Synonyms: CXCL13; SCYB13; BCA1; BLR1L; ANGIE; ANGIE2; Chemokine (C-X-C-Motif) ligand 13; Small Inducible Cytokine B Subfamily (Cys-X-Cys Motif) Member 13. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7228