Learn More
Raybiotech Inc Mouse BMP-9 ELISA Kit
Supplier:  Raybiotech Inc ELMBMP91
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 4 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects mouse and human BMP-9. Other species not determined.
Catalog No. 50-194-9935