Learn More
BIOHIPPO Mouse BMP1 Microsample
SDP
Catalog No. 5000315920
Encompass
Mouse BMP1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BMP1 in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 30 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse BMP1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse BMP1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse BMP1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15920 Supplier BIOHIPPO Supplier No. BHE15212165
Mouse BMP1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BMP1 in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 30 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse BMP1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse BMP1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse BMP1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color