Learn More
BIOHIPPO Mouse BMPR1A Microsample
SDP
Catalog No. 5000315934
Encompass
Mouse BMPR1A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BMPR1A in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 059 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse BMPR1A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse BMPR1A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse BMPR1A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15934 Supplier BIOHIPPO Supplier No. BHE15212245
Mouse BMPR1A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BMPR1A in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 059 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse BMPR1A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse BMPR1A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse BMPR1A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color