Learn More
Sino Biological Anti-BMPR1A/ALK-3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50078RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse ALK-3 (rM ALK-3; Catalog#50078-M08H; NP_033888.2; Met 1-Arg 152). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-7664