Learn More
Sino Biological Mouse BMPR1A PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50078RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse ALK-3 (rM ALK-3; Catalog#50078-M08H; NP_033888.2; Met 1-Arg 152). ALK-3 specific IgG was purified by mouse ALK-3 affinity chromatography.
Catalog No. 50-237-3313