Learn More
Biomatik Corporation Mouse BMX Non Receptor Tyrosine Kinase (BMX) ELISA Kit
Supplier:  Biomatik Corporation EKU10070
Encompass_Preferred
Mouse BMX Non Receptor Tyrosine Kinase (BMX) ELISA Kit for detecting BMX in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97504. Gene ID: 12169. Target Synonyms: ETK; PSCTK3; NTK38; Epithelial and endothelial tyrosine kinase; Bone marrow tyrosine kinase gene in chromosome X; Cytoplasmic tyrosine-protein kinase BMX. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7601