Learn More
American Research Products Inc Mouse Bone morphogenetic prote
Supplier:  American Research Products Inc CSBE09279M
Encompass
Mouse Bone morphogenetic protein 6 BMP-6 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-30164