Learn More
American Research Products Inc Mouse bone morphogenetic prote
Catalog No. 5000329455
Encompass
Mouse bone morphogenetic protein 9 BMP-9 ELISA Kit from CUSABIO Detection Range 0 312 pg/mL-20 pg/mL Sensitivity 0 078 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29455 Supplier American Research Products Inc Supplier No. CSBE14282M
Mouse bone morphogenetic protein 9 BMP-9 ELISA Kit from CUSABIO Detection Range 0 312 pg/mL-20 pg/mL Sensitivity 0 078 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich