Learn More
American Research Products Inc Mouse Bone morphogenetic prote
Supplier:  American Research Products Inc CSBEL002733MO
Encompass
Mouse Bone morphogenetic protein 1 (BMP1) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27424