Learn More
Biomatik Corporation Mouse Bone Morphogenetic Protein 10 (BMP10) CLIA Kit
Supplier:  Biomatik Corporation EKU09969
Encompass_Preferred
Mouse Bone Morphogenetic Protein 10 (BMP10) CLIA Kit for detecting BMP10 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9R229. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6888