Learn More
Biomatik Corporation Mouse Bone Morphogenetic Protein 3 (BMP3) ELISA Kit
Supplier:  Biomatik Corporation EKU12284
Encompass_Preferred
Mouse Bone Morphogenetic Protein 3 (BMP3) ELISA Kit for detecting BMP3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8BHE5. Gene ID: 110075. Target Synonyms: BMP3A; BMP-3A; Osteogenic. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7706