Learn More
Biomatik Corporation Mouse Bone Morphogenetic Protein 9 (BMP9) CLIA Kit
Supplier:  Biomatik Corporation EKU09695
Encompass_Preferred
Mouse Bone Morphogenetic Protein 9 (BMP9) CLIA Kit for detecting BMP9 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.10-800pg/ml. Sensitivity: 0.42pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9WV56. Gene ID: 12165. Target Synonyms: GDF2; Growth Differentiation Factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8464