Learn More
American Research Products Inc Mouse Bone morphogenetic prote
Catalog No. 5000328761
Encompass
Mouse Bone morphogenetic protein 2 BMP-2 ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28761 Supplier American Research Products Inc Supplier No. CSBE04509M
Mouse Bone morphogenetic protein 2 BMP-2 ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich