Learn More
American Research Products Inc Mouse Bone morphogenetic prote
Catalog No. 5000328868
Encompass
Mouse Bone morphogenetic protein 7 BMP-7 ELISA Kit from CUSABIO Detection Range 0 625 pg/mL-40 pg/mL Sensitivity 0 156 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28868 Supplier American Research Products Inc Supplier No. CSBE08125M
Mouse Bone morphogenetic protein 7 BMP-7 ELISA Kit from CUSABIO Detection Range 0 625 pg/mL-40 pg/mL Sensitivity 0 156 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich