Learn More
Biomatik Corporation Mouse Bone Morphogenetic Protein 7 (BMP7) ELISA Kit
Supplier:  Biomatik Corporation EKU02764
Encompass_Preferred
Mouse Bone Morphogenetic Protein 7 (BMP7) ELISA Kit for detecting BMP7 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P23359. Gene ID: 12162. Target Synonyms: OP1; Osteogenic Protein-1; Eptotermin alfa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4991