Learn More
Biomatik Corporation Mouse Bone Sialoprotein (BSP) ELISA Kit
Supplier:  Biomatik Corporation EKU02773
Encompass_Preferred
Mouse Bone Sialoprotein (BSP) ELISA Kit for detecting BSP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61711. Gene ID: 15891. Target Synonyms: IBSP; Bsp; BspII; SpII; Integrin-Binding Bone Sialoprotein; Cell-binding sialoprotein; Bone sialoprotein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4521