Learn More
Abcam Mouse Bone Sialoprotein ELISA Kit (IBSP), 1 x 96 Tests
Catalog No. 502671666
Encompass_Preferred
Mouse Bone Sialoprotein ELISA Kit (IBSP) is a single-wash 90-min Simplestep used to quantify Mouse Bone Sialoprotein (IBSP) with a sensitivity of 6 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1666 Supplier Abcam Supplier No. AB2794051X96TESTS
Mouse Bone Sialoprotein ELISA Kit (IBSP) is a single-wash 90-min Simplestep used to quantify Mouse Bone Sialoprotein (IBSP) with a sensitivity of 6 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement