Learn More
Lonza Walkersville Mouse Brain CD1 Hippoc Neurons
Catalog No. 5000496363
Encompass
Clonetics Mouse Hippopcampal Neurons are isolated from E16-17 CD-1 mice The hippocampus is a major component of the brains of humans and other mammals It belongs to the limbic system and plays important roles in long-term memory and spatial navigation Clonetics Mouse Hippocampal Neurons are cryopreserved at primary passage They stain positive for PGP and Tuj-1 and test negative for sterility and mycoplasma Cells are guaranteed when using Clonetics PNGM media and contain 1M cells per vial Recommended medium PNGM BulletKit (CC-4461)
Catalog No. 50-004-96363 Supplier Lonza Walkersville Supplier No. MHI401
Clonetics Mouse Hippopcampal Neurons are isolated from E16-17 CD-1 mice The hippocampus is a major component of the brains of humans and other mammals It belongs to the limbic system and plays important roles in long-term memory and spatial navigation Clonetics Mouse Hippocampal Neurons are cryopreserved at primary passage They stain positive for PGP and Tuj-1 and test negative for sterility and mycoplasma Cells are guaranteed when using Clonetics PNGM media and contain 1M cells per vial Recommended medium PNGM BulletKit (CC-4461)