Learn More
Biomatik Corporation Mouse Brain Finger Protein (BFP) ELISA Kit
Supplier:  Biomatik Corporation EKU02789
Encompass_Preferred
Mouse Brain Finger Protein (BFP) ELISA Kit for detecting BFP in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.066ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96DY5. Gene ID: 22671. Target Synonyms: RNF179; RNF112; Ring Finger Protein 179; Ring Finger Protein 112. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6048